missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC5A/RAI3 Antibody (GPRC5A/7893) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24315JF635
This item is not returnable.
View return policy
Description
GPRC5A/RAI3 Monoclonal antibody specifically detects GPRC5A/RAI3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| GPRC5A/RAI3 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| G protein-coupled receptor, family C, group 5, member A, GPCR5A, Orphan G-protein-coupling receptor PEIG-1, RAI3G-protein coupled receptor family C group 5 member A, RAIG1RAIG-1, retinoic acid induced 3, retinoic acid responsive, Retinoic acid-induced gene 1 protein, retinoic acid-induced protein 3 | |
| Recombinant fragment (around aa200-357) of human GPRC5A/RAI3 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| GPCR | |
| 9052 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| GPRC5A/7893 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction