missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC5A/RAI3 Antibody (481906), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5239D
This item is not returnable.
View return policy
Description
GPRC5A/RAI3 Monoclonal antibody specifically detects GPRC5A/RAI3 in Human samples. It is validated for Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| GPRC5A/RAI3 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 481906 | |
| Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor, family C, group 5, member A, GPCR5A, Orphan G-protein-coupling receptor PEIG-1, RAI3G-protein coupled receptor family C group 5 member A, RAIG1RAIG-1, retinoic acid induced 3, retinoic acid responsive, Retinoic acid-induced gene 1 protein, retinoic acid-induced protein 3 | |
| NS0 mouse myeloma cell line transfected with human GPRC5A, Met1-Ser357, Accession # Q8NFJ5 | |
| 0.1 mL | |
| GPCR | |
| 9052 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction