missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRASP1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06130CL1
This item is not returnable.
View return policy
Description
GPRASP1 Polyclonal antibody specifically detects GPRASP1 in Human samples. It is validated for Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| GPRASP1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| G protein-coupled receptor associated sorting protein 1, GASP1, GASPKIAA0443GASP-1, G-protein coupled receptor-associated sorting protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human GPRASP1 fragment. | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 9737 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction