Learn More
GPR97 Antibody (CL11474) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21195AF647
Product Code. 30300312
  • £558.00 / 0.10mL
Description
GPR97 Monoclonal antibody specifically detects GPR97 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR97
Monoclonal
Alexa Fluor 647
G protein-coupled receptor 97, Pb99, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97
This antibody was developed using a recombinant protein derived from Q86Y34, with the exact immunogen sequence remaining proprietary.
0.1 mL
GPCR
222487
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11474
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified