missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR97 Antibody (CL11463) [Alexa Fluor™ 750], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21194AF750
This item is not returnable.
View return policy
Description
GPR97 Monoclonal antibody specifically detects GPR97 in Human samples. It is validated for Western Blot
Specifications
| GPR97 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| G protein-coupled receptor 97, Pb99, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97 | |
| This antibody was developed using a synthetic peptide derived from Q86Y34, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| GPCR | |
| 222487 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot | |
| CL11463 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido