missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR89B Monoclonal Antibody (4D8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA521938
This item is not returnable.
View return policy
Description
Peptide Sequence: SYFLRNVTDT DILALERRLL QTMDMIISKK KRMAMARRTM FQKGEVHNKP SGFWGMIKSV TTSASGSENL TLIQQEVDAL EELSRQLFLE TADLYATKER IEYSKTFKGK.
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
Specifications
| GPR89B | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| P0CG08 | |
| GPR89B | |
| GPR89B (AAH03187, 175 a.a. ~ 284 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA | |
| 4D8 | |
| Unconjugated | |
| GPR89B | |
| CGI-13; G protein-coupled receptor 89B; G protein-coupled receptor 89C; Golgi pH regulator A; Golgi pH regulator B; GPHR; GPHRA; GPHRB; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; protein GPR89; Protein GPR89A; Protein GPR89B; putative Golgi pH regulator C; putative MAPK-activating protein PM01; Putative NF-kappa-B-activating protein 90; putative protein GPR89C; SH120; UNQ192; UNQ192/PRO218 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51463 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction