missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR89A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118817
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total GPR89A.
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
Specifications
| GPR89A | |
| Polyclonal | |
| Unconjugated | |
| GPR89A | |
| 4933412D19Rik; CGI-13; G protein-coupled receptor 89; G protein-coupled receptor 89A; Golgi pH regulator; Golgi pH regulator A; Golgi pH regulator B; Gphr; GPHRA; GPHRB; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; Protein GPR89; Protein GPR89A; Protein GPR89B; Putative MAPK-activating protein PM01; putative NF-kappa-B-activating protein 90; SH120; STRG; UNQ192; UNQ192/PRO218 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 653519, 67549 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B7ZAQ6, Q8BS95 | |
| Gpr89, GPR89A | |
| A synthesized peptide derived from human GPR89A(Accession B7ZAQ6), corresponding to amino acid residues Q248-V298. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction