missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR89A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533785
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Mouse, Rat, Dog, Bat, Bovine, Hamster, Panda, Pig, Chicken, Platypus, Xenopus (100%).
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
Specifications
| GPR89A | |
| Polyclonal | |
| Unconjugated | |
| GPR89A | |
| 4933412D19Rik; CGI-13; G protein-coupled receptor 89; G protein-coupled receptor 89A; Golgi pH regulator; Golgi pH regulator A; Golgi pH regulator B; Gphr; GPHRA; GPHRB; GPR89; GPR89A; GPR89B; GPR89C; HSPC201; Protein GPR89; Protein GPR89A; Protein GPR89B; Putative MAPK-activating protein PM01; putative NF-kappa-B-activating protein 90; SH120; STRG; UNQ192; UNQ192/PRO218 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 483157, 653519, 67549, 767859 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| B7ZAQ6, Q5BIM9, Q8BS95 | |
| Gpr89, GPR89A | |
| Synthetic 18 amino acid peptide from 3rd cytoplasmic domain of human GPR89A. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine, Hamster, Pig, Chicken, Frog, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction