missing translation for 'onlineSavingsMsg'
Learn More

GPR89 Antibody (4D8), Novus Biologicals™

Product Code. 18367589 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18367589 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18367589 Supplier Novus Biologicals Supplier No. H00051463M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GPR89 Monoclonal antibody specifically detects GPR89 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR89
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4D8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH03187
Gene Alias FLJ55546, G protein-coupled receptor 89, G protein-coupled receptor 89B, Golgi pH regulator A, Golgi pH regulator B, GPHR, GPHRB, GPR89, protein GPR89, Protein GPR89B, putative MAPK-activating protein PM01, putative NF-kappa-B-activating protein 90, SH120
Host Species Mouse
Immunogen GPR89 (AAH03187, 175 a.a. ~ 284 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. SYFLRNVTDTDILALERRLLQTMDMIISKKKRMAMARRTMFQKGEVHNKPSGFWGMIKSVTTSASGSENLTLIQQEVDALEELSRQLFLETADLYATKERIEYSKTFKGK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51463
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.