missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR88 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533783
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Bovine (100%) Rat (94%) Mouse, Rabbit, Opossum (89%).
GPR88 is an Orphan-A GPCR with an unknown ligand. GPR88 expression has been documented almost exclusively in the striatum of the brain and not in any peripheral tissues. ESTs have been isolated from diseased brain, ear, and kidney libraries.
Specifications
| GPR88 | |
| Polyclonal | |
| Unconjugated | |
| GPR88 | |
| AW061286; COCPMR; G protein coupled receptor 88; G protein-coupled receptor 88; Gpr88; G-protein coupled receptor 88; Probable G-protein coupled receptor 88; Strg; Striatum-specific G-protein coupled receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54112, 781302 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9GZN0 | |
| GPR88 | |
| Synthetic 18 amino acid peptide from 2nd cytoplasmic domain of human GPR88. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction