missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR88 Polyclonal Antibody

GPR88 Polyclonal Antibody
Brand: Invitrogen PA533782
This item is not returnable.
View return policy
Description
GPR88 Polyclonal Antibody for IHC (P)
Specifications
| GPR88 | |
| Polyclonal | |
| Unconjugated | |
| GPR88 | |
| AW061286; COCPMR; G protein coupled receptor 88; G protein-coupled receptor 88; Gpr88; G-protein coupled receptor 88; Probable G-protein coupled receptor 88; Strg; Striatum-specific G-protein coupled receptor | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100346034, 100623311, 54112, 64378, 64443 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9EPB7, Q9ESP4, Q9GZN0 | |
| GPR88 | |
| Synthetic 19 amino acid peptide from C-terminus of human GPR88. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Monkey, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction