missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR88 Antibody (CL15190) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
£321.00 - £459.00
Specifications
| Antigen | GPR88 |
|---|---|
| Clone | CL15190 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279456
|
Novus Biologicals
NBP3-43656-100ul |
100 μL |
£459.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279550
|
Novus Biologicals
NBP3-43656-25ul |
25 μL |
£321.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR88 Monoclonal antibody specifically detects GPR88 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| GPR88 | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| GPCR | |
| PBS, pH 7.2, 40% glycerol | |
| 54112 | |
| IgG1 | |
| Protein A purified |
| CL15190 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:20-1:50, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| G protein coupled receptor 88, G protein-coupled receptor 88, G-protein coupled receptor 88, probable G-protein coupled receptor 88, STRG, Striatum-specific G-protein coupled receptor | |
| This antibody was generated using a synthetic peptide of Q9GZN0, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title