missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GPR88 Polyclonal antibody specifically detects GPR88 in Human, Mouse, Rat, Bovine, Hamster, Primate, Monkey, Rabbit samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | GPR88 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry 10-20 μg/mL, Immunohistochemistry-Paraffin 10-20 μg/mL |
| Formulation | PBS |
| Gene Accession No. | Q9GZN0 |
| Gene Alias | G protein coupled receptor 88, G protein-coupled receptor 88, G-protein coupled receptor 88, probable G-protein coupled receptor 88, STRG, Striatum-specific G-protein coupled receptor |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?