Learn More
Invitrogen™ GPR87 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA3043
Description
IHC(P) analysis shows positive staining of GPR87 in human kidney tubules. WB analysis shows GPR87 in glyco-protein enriched fractions from GPR84 or GPR87 overexpressed 293 cells. The GPR87 antiserum showed specific detection of GPR87 over other tested GPRs. GPR87 exists in different glycosylated forms.
G protein-coupled receptors play a role in cell communication. They are characterized by an extracellular N terminus, 7 transmembrane regions, and an intracellular C terminus. GPR87 has been reported to be expressed in human prostate and rat stomach and fetal tissues. ESTs have been isolated from B-cell/lung/testis, breast, heart, jaw, prostate, and testis libraries.
Specifications
| GPR87 | |
| Polyclonal | |
| Unconjugated | |
| GPR87 | |
| FKSG78; G protein-coupled receptor 87; G protein-coupled receptor 95; GPR87; GPR95; G-protein coupled receptor 87; G-protein coupled receptor 95; KPG_002; MGC131898; orphan GPCR 87 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Conc. Not Determined | |
| whole serum with 0.05% sodium azide | |
| Q9BY21 | |
| GPR87 | |
| KLH conjugated COOH-terminal amino acids of 339-358 (RSLQSVRRSEVRIYYDYTDV). | |
| RUO | |
| 53836 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.