missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR87 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533780

Product Code. 15312481

  • £439.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gibbon, Monkey, Marmoset, Bovine, Dog, Bat, Hamster, Elephant, Panda, Horse, Rabbit, Pig, Opossum, Platypus (100%).

G protein-coupled receptors play a role in cell communication. They are characterized by an extracellular N terminus, 7 transmembrane regions, and an intracellular C terminus. GPR87 has been reported to be expressed in human prostate and rat stomach and fetal tissues. ESTs have been isolated from B-cell/lung/testis, breast, heart, jaw, prostate, and testis libraries.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPR87
Polyclonal
Unconjugated
GPR87
FKSG78; G protein-coupled receptor 87; G protein-coupled receptor 95; GPR87; GPR95; G-protein coupled receptor 87; G-protein coupled receptor 95; KPG_002; MGC131898; orphan GPCR 87
Rabbit
Antigen Affinity Chromatography
RUO
100050286, 100344243, 100524399, 100770345, 485718, 53836, 785030
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q9BY21
GPR87
Synthetic 19 amino acid peptide from C-terminus of human GPR87.
50 μg
Primary
Human, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.