Learn More
Invitrogen™ GPR85 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA599893
Product Code. 16694135
  • £448.00 / 100µL
Description
Antibody detects endogenous levels of total GPR85.
GPR85, a member of the SREB (Super-Conserved Receptor Expressed in Brain) family of GPCRs, is an Orphan-A GPCR with an unknown ligand. The rat counterpart of SREB2 is involved in the development of the cerebral cortex. GPR85 has been reported to be expressed in several areas of human and rat CNS (Matsumoto et al., 2000), as well as in genital organs. ESTs have been isolated from B-cell/lung/testis, brain, testis, and embryo libraries, as well as many mouse tissue libraries.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GPR85
Polyclonal
Unconjugated
GPR85
2900026B03Rik; FLJ50993; FLJ51159; FLJ99433; G protein-coupled receptor 85; Gpr85; PKrCx1; probable G-protein coupled receptor 85; Rgpr85; seven transmembrane helix receptor; SREB; SREB2; Srep2; Super conserved receptor expressed in brain 2
Rabbit
Affinity Chromatography
RUO
54329, 64020, 64450
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P60893, P60894, P60895
GPR85
A synthesized peptide derived from human GPR85(Accession P60893), corresponding to amino acid residues G186-S236.
100 μL
Primary
Human, Mouse, Rat