missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR83 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23850-1-AP-20UL
This item is not returnable.
View return policy
Description
GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.Specifications
| GPR83 | |
| Polyclonal | |
| Unconjugated | |
| GPR83 | |
| G protein coupled receptor 72, G protein coupled receptor 83, GIR, GPR72, GPR83, KIAA1540 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10888 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9NYM4 | |
| GPR83 | |
| GPR83 Fusion Protein Ag20873 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction