missing translation for 'onlineSavingsMsg'
Learn More

GPR83 Antibody, Biotin, Novus Biologicals™

Product Code. 18402332
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402332 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402332 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224515B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR83 Polyclonal antibody specifically detects GPR83 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR83
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias G protein-coupled receptor 72, G protein-coupled receptor 83, GIR, GPR72glucocorticoid induced receptor, G-protein coupled receptor 72, KIAA1540, probable G-protein coupled receptor 83
Host Species Rabbit
Immunogen A portion of amino acids 200-250 of human GPR83 was used as the immunogen for this antibody.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 10888
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.