missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR83 Antibody (566133), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB10943MFV500
This item is not returnable.
View return policy
Description
GPR83 Monoclonal antibody specifically detects GPR83 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR83 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 566133 | |
| Flow Cytometry | |
| G protein-coupled receptor 72, G protein-coupled receptor 83, GIR, GPR72glucocorticoid induced receptor, G-protein coupled receptor 72, KIAA1540, probable G-protein coupled receptor 83 | |
| NS0 mouse myeloma cell line transfected with human GPR83, Accession # Q9NYM4 | |
| 0.1 mL | |
| GPCR | |
| 10888 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction