missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR81 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519144
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
The G protein-coupled receptor GPCR GPR81 has been reported primarily in adipose and pituitary. ESTs have been isolated from human bladder cancer and colon cancer libraries. It has not been detected in frontal, temporal and occipital lobes of the cortex, basal forebrain, caudate nucleus, nucleus accumbens, and hippocampus.
Specifications
| GPR81 | |
| Polyclonal | |
| Unconjugated | |
| HCAR1 | |
| FKSG80; G protein-coupled receptor 104; G protein-coupled receptor 81; GPR104; GPR81; G-protein coupled receptor 104; G-protein coupled receptor 81; HCA1; HCAR1; hydrocarboxylic acid receptor 1; hydroxycarboxylic acid receptor 1; hydroxy-carboxylic acid receptor 1; LACR1; lactate receptor 1; TAGPCR; TA-GPCR; T-cell activation G protein-coupled receptor | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 27198 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9BXC0 | |
| HCAR1 | |
| Synthetic peptide sequence (QQLARQARMKKATR) corresponding to the internal amino acids of GPR81 (aa 207-221). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction