missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR81 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-51956
This item is not returnable.
View return policy
Description
GPR81 Polyclonal specifically detects GPR81 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| GPR81 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FKSG80, G protein-coupled receptor 104, G protein-coupled receptor 81, GPR104, G-protein coupled receptor 104, G-protein coupled receptor 81, HCA1, HCAR1, LACR1, lactate receptor 1, TA-GPCR, T-cell activation G protein-coupled receptor | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Expected from sequence similarity: Mouse, Rat, Dog, Pig and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:8000 | |
| NP_115943.1 | |
| HCAR1 | |
| Peptide with sequence C-QQLARQARMKKATR corresponding to internal region according to NP_115943.1. | |
| 0.1 mg | |
| Chemokine Research, GPCR, Immunology | |
| 27198 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction