missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR78 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533764
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee (100%) Gorilla, Orangutan, Gibbon (94%) Monkey (88%).
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
Specifications
| GPR78 | |
| Polyclonal | |
| Unconjugated | |
| GPR78 | |
| G protein-coupled receptor 78; GPR78; G-protein coupled receptor 78; UNQ5925/PRO19818 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q96P69 | |
| GPR78 | |
| Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human GPR78. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction