missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR77 Monoclonal Antibody (6C1G10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA548731
This item is not returnable.
View return policy
Description
GPR77 Monoclonal Antibody for ELISA, Flow, ICC/IF, IHC (P), Western Blot
C5AR2 is an orphan member that belongs to anaphylatoxin receptor family. It is a seven transmembrane G-protein-coupled receptor consisting of an N-terminal N-glycosylation site, several conserved cysteines and several potential C-terminal phosphorylation sites. It binds to C5a and C5a-des-Arg and serves as a highly regulated scavenger receptor with a negative modulatory influence on C5aR-mediated inflammation. Northern Blot analysis suggests it is co- expressed along with C5aR in different tissues and cells including neutrophils/macrophages, mast cells, dendritic cells, brain, lung, heart, kidney, liver, ovary and testis.
Specifications
| GPR77 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q9P296 | |
| C5AR2 | |
| Purified recombinant fragment of human C5AR2 (AA: extra mix) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 6C1G10 | |
| Unconjugated | |
| C5AR2 | |
| C5a anaphylatoxin chemotactic receptor 2; C5a anaphylatoxin chemotactic receptor C5L2; C5AR2; C5L2; Complement component 5a receptor 2; G protein-coupled receptor 77; G protein-coupled receptor C5L2; GPF77; GPR77; G-protein coupled receptor 77 | |
| Mouse | |
| Protein G | |
| RUO | |
| 27202 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction