missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR75 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567870
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
GPR75 Polyclonal antibody specifically detects GPR75 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| GPR75 | |
| Polyclonal | |
| Unconjugated | |
| GPR75 | |
| G protein-coupled receptor 75; GPR75; GPRchr2; GPR-chr2; Probable G-protein coupled receptor 75; WI31133; WI-31133 | |
| Rabbit | |
| affinity chromatography | |
| RUO | |
| 10936, 237716 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95800, Q6X632 | |
| GPR75 | |
| Synthesized peptide derived from human GPR75. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction