missing translation for 'onlineSavingsMsg'
Learn More

GPR74/NPFFR2 Antibody, Novus Biologicals™

Product Code. 18254494 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18254494 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18254494 Supplier Novus Biologicals Supplier No. NBP169056

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR74/NPFFR2 Polyclonal specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR74/NPFFR2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. A0PJM9
Gene Alias GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77
Gene Symbols NPFFR2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to NPFFR2 (neuropeptide FF receptor 2) The peptide sequence was selected from the C terminal of NPFFR2. Peptide sequence KAKSHVLINTSNQLVQESTFQNPHGETLLYRKSAEKPQQELVMEELKETT.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 10886
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.