missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
GPR74/NPFFR2 Polyclonal antibody specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Host Species | Rabbit |
| Immunogen | Reacts with the human NPFF2 protein. |
| Purification Method | Immunogen affinity purified |
| Regulatory Status | RUO |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?