missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR74/NPFFR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £309.00
Specifications
| Antigen | GPR74/NPFFR2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18107012
|
Novus Biologicals
NBP2-24879SS |
0.025 mg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18101813
|
Novus Biologicals
NBP2-24879 |
0.1 mg |
£309.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GPR74/NPFFR2 Polyclonal specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot.Specifications
| GPR74/NPFFR2 | |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| PBS with 0.02% Sodium Azide | |
| 10886 | |
| Reacts with the human NPFF2 protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77 | |
| NPFFR2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title