Learn More
GPR65 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20306-1-AP-20UL
This item is not returnable.
View return policy
Description
The GPR65 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human GPR65. This antibody recognizes human antigen. The GPR65 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| GPR65 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 8477 | |
| IgG |
| Immunohistochemistry, ELISA | |
| 393 μg/mL | |
| Rabbit | |
| Primary | |
| Human |