missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR64 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533752
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Dog, Bat, Pig (100%) Marmoset, Bovine, Panda, Horse (94%) Elephant, Rabbit (88%).
Could be involved in a signal transduction pathway controlling epididymal function and male fertility.Specifications
GPR64 | |
Polyclonal | |
Unconjugated | |
ADGRG2 | |
ADGRG2; adhesion G protein-coupled receptor G2; adhesion G-protein coupled receptor G2; AW212196; B830041D06Rik; CTD-2245E12.1; EDDM6; epididymal protein 6; epididymis-specific protein 6; FLJ00282; G protein-coupled receptor 64; G protein-coupled receptor, epididymis-specific (seven transmembrane family); Gpr64; G-protein coupled receptor 64; He6; Human epididymis-specific protein 6; LNB-TM7 heptahelical receptor Me6; Me6; MGC104454; MGC138738; MGC138739; Mouse epididymis-specific protein 6; Rat epididymis-specific protein 6; Re6; re6 receptor; TM7LN2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100157214, 10149, 491763 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8IZP9 | |
ADGRG2 | |
Synthetic 16 amino acid peptide from C-terminus of human GPR64. | |
50 μg | |
Primary | |
Canine, Human, Porcine | |
Antibody | |
IgG |