missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR64 Antibody (864238), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB79771APCCY7
This item is not returnable.
View return policy
Description
GPR64 Monoclonal antibody specifically detects GPR64 in Human samples. It is validated for Flow Cytometry
Specifications
| GPR64 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 864238 | |
| Flow Cytometry | |
| EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, MGC104454, MGC138738, TM7LN2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human GPR64, Leu38-Asn64, Glu68-Thr553, Accession # NP_001073328 | |
| 0.1 mL | |
| GPCR | |
| 10149 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction