missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR63 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111106
This item is not returnable.
View return policy
Description
Immunogen sequence: NTTFVVYENT YMNITLPPPF QHPDLSPLLR YSFETMAPTG LSSLTVNSTA VPTTPAAFKS LNLPLQ Highest antigen sequence indentity to the following orthologs: Mouse - 68%, Rat - 71%.
G protein-coupled receptors (GPCRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPR63 is a member of this family (subfamily: lysophospholipid/lysosphingolipid) and its expression has been reported in several brain regions, including caudate, frontal cortex, hypothalamus, midbrain, and thalamus, as well as in small intestine, stomach, and thymus.
Specifications
| GPR63 | |
| Polyclonal | |
| Unconjugated | |
| GPR63 | |
| brain expressed G-protein-coupled receptor PSP24 beta; G protein-coupled receptor 63; G protein-coupled receptor PSP24-2; Gpr63; Probable G-protein coupled receptor 63; PSP24(beta); PSP24-2; PSP24B; PSP24beta; PSP24-beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81491 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BZJ6 | |
| GPR63 | |
| Recombinant Protein corresonding to Human GPR63. Recombinant protein control fragment (Product #RP-95306). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction