Learn More
Invitrogen™ GPR61 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532832
Description
Percent identity with other species by BLAST analysis: Human, Monkey, Marmoset, Elephant, Panda (100%); Gorilla, Gibbon, Mouse, Rat, Bat, Bovine, Hamster, Horse, Pig, Opossum (95%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR61 is a member of this family (subfamily Orphan-A). It has been reported to be expressed in several regions of brain, including caudate, putamen, and thalamus.
Specifications
| GPR61 | |
| Polyclonal | |
| Unconjugated | |
| GPR61 | |
| BALGR; biogenic amine receptor-like GPCR; Biogenic amine receptor-like G-protein coupled receptor; G protein-coupled receptor 61; GPCR3; GPR61; G-protein coupled receptor 61; probable G-protein coupled receptor 61 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 83873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9BZJ8 | |
| GPR61 | |
| Synthetic 19 amino acid peptide from C-terminus of human GPR61. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.