missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR52 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533734
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon (95%) Panda (90%) Mouse, Bat, Bovine (85%) Rabbit (80%).
GPR52 is a member of the G protein-coupled receptor (GPR) family play important roles in signal transduction from the external environment to the inside of the cell.
Specifications
| GPR52 | |
| Polyclonal | |
| Unconjugated | |
| GPR52 | |
| Axor10; EG620246; G protein-coupled receptor 52; Gpr52; G-protein coupled receptor 52; MGC111751; probable G-protein coupled receptor 52 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9293 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9Y2T5 | |
| GPR52 | |
| Synthetic 20 amino acid peptide from N-terminal extracellular domain of human GPR52. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction