missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GPR52 Comp-43

Description
GPR52 Comp-43 is a GPR52 receptor antagonist (IC50 = 0.63 μM). The use of GPR52 Comp-43 in animal models of Huntington's Disease leads to degradation and reduction of soluble and aggregated mutant huntingtin protein in the striatum. It also reduces neuronal loss, promotes neuronal survival and improves motor functions in vivo. GPR52 Comp-43 is active in vitro and brain penetrant.
Specifications
Specifications
| Chemical Name or Material | (2 E)-5-Phenyl-1-(2-thienyl)-2-penten-1-one |
| CAS | 1239987-91-7 |
| Quantity | 10 mg |
| Target | Orphan 7-TM Receptor Antagonists |
| Molecular Formula | C15H14OS |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?