missing translation for 'onlineSavingsMsg'
Learn More

GPR50 Antibody, Novus Biologicals™

Product Code. 18377488 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377488 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377488 Supplier Novus Biologicals Supplier No. H00009248B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

GPR50 Polyclonal antibody specifically detects GPR50 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR50
Applications Western Blot, Flow Cytometry, KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry, Knockout Validated
Formulation PBS (pH 7.4)
Gene Accession No. AAI03697.1
Gene Alias G protein-coupled receptor 50H9Mel1c, melatonin-related receptor, MGC125342
Host Species Mouse
Immunogen GPR50 (AAI03697.1, 1 a.a. - 617 a.a.) full-length human protein. MGPTLAVPTPYGCIGCKLPQPEYPPALIIFMFCAMVITIVVDLIGNSMVILAVTKNKKLRNSGNIFVVSLSVADMLVAIYPYPLMLHAMSIGGWDLSQLQCQMVGFITGLSVVGSIFNIVAIAINRYCYICHSLQYERIFSVRNTCIYLVITWIMTVLAVLPNMYIGTIEYDPRTYTCIFNYLNNPVFTVTIVCIHFVLPLLIVGFCYVRIWTKVLAARDPAGQNPDNQLAEVRNFLTMFVIFLLFAVCWCPINVLTVLVAVSPKEMAGKIPNWLYLAAYFIAYFNSCLNAVIYGLLNENFRREYWTIFHAMRHPIIFFSGLISDIREMQEARTLARARAHARDQAREQDRAHACPAVEETPMNVRNVPLPGDAAAGHPDRASGHPKPHSRSSSAYRKSASTHHKSVFSHSKAASGHLKPVSGHSKPASGHPKSATVYPKPASVHFKADSVHFKGDSVHFKPDSVHFKPASSNPKPITGHHVSAGSHSKSAFSAATSHPKPTTGHIKPATSHAEPTTADYPKPATTSHPKPTAADNPELSASHCPEIPAIAHPVSDDSDLPESASSPAAGPTKPAASQLESDTIADLPDPTVVTTSTNDYHDVVVVDVEDDPDEMAV
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9248
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.