missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR45 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533724
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset, Bovine, Panda (100%) Galago, Elephant, Rabbit, Horse, Pig (94%).
This intronless gene encodes a member of the G protein-coupled receptor (GPCR) family. Members of this protein family contain seven putative transmembrane domains and may mediate signaling processes to the interior of the cell via activation of heterotrimeric G proteins. This protein may function in the central nervous system.
Specifications
| GPR45 | |
| Polyclonal | |
| Unconjugated | |
| GPR45 | |
| 9230112G11Rik; AW046772; G protein-coupled receptor 45; GPR45; high-affinity lysophosphatidic acid receptor; probable G-protein coupled receptor 45; PSP24; PSP24(ALPHA); PSP24-1; PSP24A; PSP24alpha; PSP24-alpha | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11250, 510657 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9Y5Y3 | |
| GPR45 | |
| Synthetic 17 amino acid peptide from C-terminus of human GPR45. | |
| 50 μg | |
| Primary | |
| Human, Monkey, Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction