missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR43 Recombinant Rabbit Monoclonal Antibody (HL3073)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556729
This item is not returnable.
View return policy
Description
GPR43 Recombinant Monoclonal Antibody for IHC (P)
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR43 is a member of this family (subfamily carboxylic acid). It expression has been reported primarily in immune cells including monocytes and neutrophils. An EST has been isolated from a human pancreatic library.
Specifications
| GPR43 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O15552 | |
| FFAR2 | |
| Recombinant protein encompassing a sequence within the Intracellular domain of human GPR43. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| HL3073 | |
| Unconjugated | |
| FFAR2 | |
| fatty acid receptor 2; FFA2; FFA2R; FFAR2; free fatty acid activated receptor 2; free fatty acid receptor 2; G protein-coupled receptor 43; GPCR43; Gpr43; G-protein coupled receptor 43; leukocyte-specific STAT-induced GPCR; Lssig | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2867 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction