missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR40 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518381
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
Specifications
| GPR40 | |
| Polyclonal | |
| Unconjugated | |
| FFAR1 | |
| FFA1R; FFAR; Ffar1; Free fatty acid receptor 1; G protein-coupled receptor 40; G protein-coupled receptor GPR40; GPCR40; GPR40; G-protein coupled receptor 40; HGNC:4498 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 2864 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O14842 | |
| FFAR1 | |
| Synthetic peptide sequence (CAARTQGGKSQK) corresponding to the C-terminus amino acids of FFAR1 (aa 289-300). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction