missing translation for 'onlineSavingsMsg'
Learn More

GPR40 Activator 1, MedChemExpress

Product Code. 16498567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498567 5 mg 5mg
16405948 10 mg 10mg
16402097 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498567 Supplier Medchem Express Supplier No. HY13971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GPR40 Activator 1 is a potent GPR40 activator for treatment of type 2 diabetes.

GPR40 Activator 1 is a potent GPR40 activator for treatment of type 2 diabetes.

Specifications

Chemical Name or Material GPR40 Activator 1
CAS 1309435-60-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C31H31NO3S
Quantity 5 mg
SMILES OC(C[C@H](C#CC)C(C=C1)=CC=C1OCC2=CC=C(CN3CCC4(C=CC5=C4C=CC=C5)CC3)S2)=O
Molecular Weight (g/mol) 497.65
Formula Weight 497.65
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.