missing translation for 'onlineSavingsMsg'
Learn More

GPR39 Antibody - BSA Free, Novus Biologicals™

Product Code. 18357554 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
Unit Size:
0.05mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18357554 0.025 mL 25µL
18313485 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18357554 Supplier Novus Biologicals Supplier No. NLS142SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 3 publications

GPR39 Polyclonal specifically detects GPR39 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR39
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. O43194, Q5U431
Gene Alias G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541
Gene Symbols GPR39
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human GPR39 protein (within residues 400-500). [Swiss-Prot O43194]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline GPCR, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2863
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.