missing translation for 'onlineSavingsMsg'
Learn More

GPR39 Antibody, Biotin, Novus Biologicals™

Product Code. 18388564 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18388564 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18388564 Supplier Novus Biologicals Supplier No. NLS139B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GPR39 Polyclonal specifically detects GPR39 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GPR39
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:100, Immunoblotting
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. O43194
Gene Alias G protein-coupled receptor 39, G-protein coupled receptor 39, MGC149541
Gene Symbols GPR39
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human GPR39 protein (within residues 150-200). [Swiss-Prot O43194]
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2863
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.