missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GPR37L1 Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1 M Urea, pH 7.4. |
| Gene ID (Entrez) | 9283 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 23 kDa |
| Quantity | 0.1 mL |
| Storage Requirements | Store at -20°C. Avoid freeze/thaw cycles. |
| Gene Alias | endothelin B receptor-like protein 2, ET(B)R-LP-2, ETBRLP2, ETBR-LP-2 endothelin type b receptor-like protein 2, G protein-coupled receptor 37 like 1, G-protein coupled receptor 37 like 1, G-protein coupled receptor 37-like 1 |
| Gene Symbol | GPR37L1 |
| Inhibitor Properties | This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-49419. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL |
| Research Category | GPCR, Neuroscience, Signal Transduction |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction