Learn More
Invitrogen™ GPR37L1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532828
Description
Percent identity with other species by BLAST analysis: Gorilla, Gibbon (100%); Human, Monkey, Marmoset (94%); Hamster (89%); Mouse, Rat (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
This polyclonal antibody was raised in Rabbit against GPR37L1. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPR37L1. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Specifications
| GPR37L1 | |
| Polyclonal | |
| Unconjugated | |
| GPR37L1 | |
| AW047233; CAG-18; D0Kist8; Endothelin B receptor-like protein 2; endothelin B receptor-like protein-2; endothelin type b receptor-like protein 2; ET(B)R-LP-2; ETBRLP2; ETBR-LP-2; G protein-coupled receptor 37 like 1; G protein-coupled receptor 37-like 1; G protein-coupled receptor 37-lke 1; GPCR/CNS2; GPR37L1; G-protein coupled receptor 37 like 1; G-protein coupled receptor 37-like 1; orphan Gpr37-like protein 1; prosaposin receptor GPR37L1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O60883 | |
| GPR37L1 | |
| Synthetic 18 amino acid peptide from N-terminal extracellular domain of human GPR37L1. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.