missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR37L1 Antibody (444532), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4449J
This item is not returnable.
View return policy
Description
GPR37L1 Monoclonal antibody specifically detects GPR37L1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| GPR37L1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 444532 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| endothelin B receptor-like protein 2, ET(B)R-LP-2, ETBRLP2, ETBR-LP-2endothelin type b receptor-like protein 2, G protein-coupled receptor 37 like 1, G-protein coupled receptor 37 like 1, G-protein coupled receptor 37-like 1 | |
| NS0 mouse myeloma cell line transfected with human GPR37L1, Accession # O60883 | |
| 0.1 mL | |
| GPCR, Neuroscience, Signal Transduction | |
| 9283 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction