missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR37 Monoclonal Antibody (436923)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561440
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Parkinson is the second most common neurodegenerative disease after Alzheimers. About 1 percent of people over the age of 65 and 3 percent of people over the age of 75 are affected by the disease. The mutation is the most common cause of Parkinson disease identified to date. The function of Park2 is not well-known; however, it may play a role in the ubiquitin-mediated proteolytic pathway. Mutations in this gene are known to cause autosomal recessive juvenile parkinsonism. Alternative splicing of this gene produces three known products of undetermined function. Panneuronal expression of Parkin substrate Pael-R causes age-dependent selective degeneration of Drosophila dopaminergic (DA) neurons; coexpression of Parkin degrades Pael-R and suppresses its toxicity.
Specifications
| GPR37 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O15354 | |
| GPR37 | |
| NS0 mouse myeloma cell line transfected with human GPR37, Ala27-Cys613. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 436923 | |
| Unconjugated | |
| GPR37 | |
| AI848630; EDNRBL; endothelin B receptor-like protein 1; ETBR-LP-1; G protein-coupled receptor 37; G protein-coupled receptor 37 (endothelin receptor type B-like); Gpcr/cns1; Gpr37; G-protein coupled receptor 37; G-protein coupled receptor CNS1; hET(B)R-LP; Het-b-r-lp; Pael receptor; PAELR; Pael-R; parkin-associated endothelin B-like receptor; Parkin-associated endothelin receptor-like receptor; probable G-protein coupled receptor 37; prosaposin receptor GPR37 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2861 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction