missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR37 Antibody (436923) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4450JF635
This item is not returnable.
View return policy
Description
GPR37 Monoclonal antibody specifically detects GPR37 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| GPR37 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| EDNRBL, Endothelin B receptor-like protein 1, G protein-coupled receptor 37 (endothelin receptor type B-like), hET(B)R-LP, PAELRETBR-LP-1, Parkin-associated endothelin receptor-like receptor, probable G-protein coupled receptor 37 | |
| NS0 mouse myeloma cell line transfected with human GPR37, Ala27-Cys613, Accession # O15354 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 436923 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| GPCR | |
| 2861 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction