missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR35 Monoclonal Antibody (C3), Invitrogen™

Mouse Monoclonal Antibody
£177.00 - £385.00
Specifications
| Antigen | GPR35 |
|---|---|
| Clone | C3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17604256
|
Invitrogen™
MA543796 |
20 μL |
£177.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17694246
![]() |
Invitrogen™
MA543795 |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Acts as a receptor for kynurenic acid, an intermediate in the tryptophan metabolic pathway. The activity of this receptor is mediated by G-proteins that elicit calcium mobilization and inositol phosphate production through G(qi/o) proteins. [UniProt]Specifications
| GPR35 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| G protein-coupled receptor 35; Gpr35; G-protein coupled receptor 35; KPG_007; KYNA receptor; kynurenic acid receptor | |
| GPR35 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| C3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9HC97 | |
| 2859 | |
| Arg240-Ala309 (Accession Q9HC97), with N-terminal His and GST Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| GPR35 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title