missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR34 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545716
This item is not returnable.
View return policy
Description
Peptide sequence: KGGHNSTMCF HYRDKHNAKG EAIFNFILVV MFWLIFLLII LSYIKIGKNL Sequence homology: Cow: 93%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 86%; Rat: 100%; Sheep: 93%.
G protein-coupled receptors (GPCRs), such as GPR34, are integral membrane proteins containing 7 putative transmembrane domains (TMs). These proteins mediate signals to the interior of the cell via activation of heterotrimeric G proteins that in turn activate various effector proteins, ultimately resulting in a physiologic response.
Specifications
| GPR34 | |
| Polyclonal | |
| Unconjugated | |
| Gpr34 | |
| G protein-coupled receptor 34; Gpr34; G-protein-coupled receptor; G-protein-coupled receptor GPR34; Lypsr1; probable G-protein coupled receptor 34 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 2857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UPC5 | |
| Gpr34 | |
| Synthetic peptide directed towards the C-terminal of human GPR34 (aa 196-245). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction