missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GPR34 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533698

Product Code. 15302471

  • £442.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BLAST analysis of the peptide immunogen showed 100% homology with human protein GPR82. Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey (100%) Marmoset, Guinea pig (95%) Mouse, Bovine, Elephant, Horse (90%).

G protein-coupled receptors (GPCRs), such as GPR34, are integral membrane proteins containing 7 putative transmembrane domains (TMs). These proteins mediate signals to the interior of the cell via activation of heterotrimeric G proteins that in turn activate various effector proteins, ultimately resulting in a physiologic response.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GPR34
Polyclonal
Unconjugated
Gpr34
G protein-coupled receptor 34; Gpr34; G-protein-coupled receptor; G-protein-coupled receptor GPR34; Lypsr1; probable G-protein coupled receptor 34
Rabbit
Antigen affinity chromatography
RUO
2857
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q9UPC5
Gpr34
Synthetic 20 amino acid peptide from C-terminus of human GPR34.
50 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.