missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR34 Monoclonal Antibody (419859)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524155
This item is not returnable.
View return policy
Description
This antibody stains human GPR34 transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
G protein-coupled receptors (GPCRs), such as GPR34, are integral membrane proteins containing 7 putative transmembrane domains (TMs). These proteins mediate signals to the interior of the cell via activation of heterotrimeric G proteins that in turn activate various effector proteins, ultimately resulting in a physiologic response.
Specifications
| GPR34 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9UPC5 | |
| Gpr34 | |
| NS0 mouse myeloma cell line transfected with human GPR34. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry | |
| 419859 | |
| Unconjugated | |
| Gpr34 | |
| G protein-coupled receptor 34; Gpr34; G-protein-coupled receptor; G-protein-coupled receptor GPR34; Lypsr1; probable G-protein coupled receptor 34 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction